Learn More
BD Affirm™ VPIII Microbial Identification Test
GSA_VA
Catalog No. 23025603 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Type:
MicroProbe™ Processor
Sample Collection Set
Sample Collection Swab
Test Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-025-603 Test Kit
23-025-600 MicroProbe™ Processor
23-025-601 Sample Collection Set
23-025-602 Sample Collection Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-025-603 Supplier BD Supplier No. 446252
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
Probe Analysis Cards (PAC) individually wrapped containing five beads (negative control, Trichomonas, Gardnerella, Candida, and positive control); and Reagent Cassettes (RC) containing patient sample reservoir, hybridization solution, wash solution, conjugate, and substrate buffer
DNA probe molecular test for the detection of vaginitis
  • Combine with BD Affirm VPIII Ambient Temperature Transport System (Cat. No. 23-025-604)
  • For detection and identification of Candida species, Gardnerella vaginalis, and Trichomonas vaginalis nucleic acid in vaginal fluid specimens from vaginitis/vaginosis patients
  • Uses two distinct single-stranded nucleic acid probes for each organism: a capture probe and color development probe complementary to unique genetic sequences of target organisms
  • Automated processing, with the BD MicroProbe™ Processor (Cat. No. 23-025-600)
  • Positive results indicated by visible blue color on target organism bead
  • Built-in positive and negative controls on each results card
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Moderate
Detectable Analytes T. vaginalis Nucleic Acid, G. vaginalis Nucleic Acid, Candida species Nucleic Acid
Type Test Kit
For Use With (Application) Detection of vaginitis
Includes Probe analysis cards individually wrapped containing five beads (negative control, Trichomonas, Gardnerella, Candida and positive control) and reagent cassettes containing patient sample reservoir, hybridization solution, wash solution, conjugateand substrate Buffer, built-in positive and negative control
Quantity 24 Tests
Test Time 2 min.