Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Anti-hamster Ig HRP Detection kit

Catalog No. BDB551012 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551012 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551012 Supplier BD Supplier No. 551012

missing translation for 'validMainframeMsg'

Includes

Biotinylated Anti-Hamster IgG Cocktail, Antibody Diluent Buffer, Streptavidin-Horseradish peroxidase (SAv-HRP), DAB Buffer, DAB Chromogen

Designed for immunohistochemical staining of acetone-fixed frozen sections, formalin- or zinc-fixed paraffin sections, and cytospins/smears

  • Used with any purified primary antibody
  • Provides maximum reactivity
  • Contains protein stabilizers to enhance antibody reactivity
  • Blocking agents reduce non-specific staining

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Storage Requirements 4°C
For Use With (Application) Immunohistochemistry, Immunohistochemical Staining
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.