Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Antibody Diluent

Catalog No. BDB559148 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559148 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559148 Supplier BD Supplier No. 559148
Add to Cart
Add to Cart

For acetone-fixed, frozen tissue sections and formalin-fixed, paraffin-embedded tissue sections

Premixed, ready to use, sodium phosphate-based buffer solution, useful for the dilution of primary and/or secondary antibodies to be used in immunohistochemistry applications

  • Storage buffer: Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody diluent
Quantity 125 mL
For Use With (Application) Immunohistochemistry (IHC), Staining of Intra-cellular Antigens
Form IHCH
Content And Storage 4°C

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.