Learn More
APC BrdU Flow Kit, 200 tests, BD
Catalog No. BDB557892 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
4 x 50 Tests
Catalog No. Quantity
BDB557892 4 x 50 Tests
1 options
Catalog No. BDB557892 Supplier BD Biosciences Supplier No. 557892
Add to Cart
Add to Cart
Comes with 65μL of APC-conjugated anti-BrdU antibody
Povides reagents for immunofluorescent BrdU staining with a protocol, compatible with the use of additional fluorescent antibodies specific for other cellular molecules. Used with fluorescent anti-cytokine antibodies in analyses of cultured cells following in vitro mitogenic stimulation of quiescent lymphoid cell populations.
  • Consists of Flow cytometer with 488nm laser capable of detecting 7-AAD and a 633- to 640nm laser capable of detecting APC
  • Staining procedure includes the fixative paraformaldehyde
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C
No. of Tests 200
Includes One 65μL vial of APC-conjugated Anti-BrdU antibody, one 25mL bottle Cytofix/Cytoperm™ buffer, two 25mL bottles perm/Wash™ buffer (10X), one 10mL bottle Cytoperm™
Quantity 4 x 50 Tests
Validated Application RUO
Product Type APC BrdU Flow Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov