Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Assay Diluent

Catalog No. BDB560104 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560104 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560104 Supplier BD Supplier No. 560104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For reconstituting and diluting standard controls and test samples

  • Can be used with Flex Set assays
  • Storage buffer: aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Diluent
Includes Storage Buffer of aqueous Buffered solution containing fetal bovine Serum and ≤0.09% Sodium Azide
Quantity 30 mL
Form Liquid

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.