Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse B Lymphocyte Activation Antibody Cocktail, with Isotype Control; PE-Cy™7 CD25, PE CD69, and APC CD19

Catalog No. BDB558063 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 Sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558063 2 Sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558063 Supplier BD Supplier No. 558063

missing translation for 'validMainframeMsg'

Includes

Mouse B Lymphocyte Activation Antibody Cocktail; PE-Cy™7 CD25, PE CD69, and APC CD19, Mouse B Lymphocyte Activation Isotype Control; PE-Cy 7, PE, and APC

Three antibodies have been titrated and pre-diluted, mixed together, and formulated for optimal staining performance. The Mouse B Lymphocyte Activation Isotype Control contains equivalent concentrations of fluorochrome- and isotype-matched negative-control immunoglobulin

  • Clone: 1D3, H1.2F3, PC61
  • Isotype: Rat IgG2a, κ, Armenian Hamster IgG1, λ3, Rat IgG1,λ
  • Permits recognition of each of the three antigens on each cell in sample
  • Expression of the three antigens identifies activated B lymphocytes
  • Additional fluorochrome-labeled reagents may be combined
  • Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 Sets

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.