Learn More
BD Mouse B Lymphocyte Activation Antibody Cocktail, with Isotype Control; PE-Cy™7 CD25, PE CD69, and FITC CD19
Shop All BD Biosciences ProductsDescription
Includes
Mouse B Lymphocyte Activation Antibody Cocktail; PE-Cy™7 CD25, PE CD69, and APC CD19, Mouse B Lymphocyte Activation Isotype Control; PE-Cy 7, PE, and APC
Three antibodies have been titrated and pre-diluted, mixed together, and formulated for optimal staining performance. The Mouse B Lymphocyte Activation Isotype Control contains equivalent concentrations of fluorochrome- and isotype-matched negative-control immunoglobulin
- Clone: 1D3, H1.2F3, PC61
- Isotype: Rat IgG2a, κ, Armenian Hamster IgG1, λ3, Rat IgG1,λ
- Permits recognition of each of the three antigens on each cell in sample
- Expression of the three antigens identifies activated B lymphocytes
- Additional fluorochrome-labeled reagents may be combined
- Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography
Flow Cytometry
Specifications
Specifications
| Quantity | 1 Set |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.