Learn More
BD BACTEC™ Bottle Plastic Plus PRIME Anaerobic Medium
Catalog No. 23025908 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
23-025-908 50/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-025-908 Supplier BD Supplier No. 442022
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pre-reduced enriched Soybean-Casein Digest broth with CO2
  • Used with BD BACTEC™ 9000 or BD BACTEC™ FX Systems
TRUSTED_SUSTAINABILITY
Specifications
Product Type Anaerobic Medium
For Use With (Equipment) BD BACTEC™9000 or BD BACTEC™ FX Systems
Quantity 50/Pk.