Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD™ Bactec™ Media: Standard/10 Aerobic/F

Catalog No. 23032512 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
50/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-032512 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-032512 Supplier BD™ Supplier No. 442260

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with BACTEC 9240, 9120, and 9050 Systems

  • Used in a qualitative procedure for the culture and recovery of anaerobic microorganisms from blood
  • Fluorescent series bar-coded blood culture media
  • Each vial contains a chemical sensor which can detect increases in CO2 produced by the growth of microorganisms
  • Vials ready to use as received—require no reconstitution or dilution
  • Detection is limited to microorganisms that will grow in a particular type of medium

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 25°C
Form Liquid
Media Type Standard/10 Aerobic/F
Formulation Processed Water 40mL, Soybean Casein digest broth 3% w/v, Yeast extract 0.3% w/v, Animal Tissue digest 0.10% w/v, Sucrose 0.1% w/v, Hemin 0.0005% w/v, Menadione 0.00005% w/v, Pyridoxal HCl (Vitamin B6) 0.001% w/v, Sodium Bicarbonate 0.04% w/v, Sodium Polyanetholsulfonate 0.035% w/v
For Use With (Equipment) Bactec™ 9240, 9120 and 9050 System
Packaging Type Vial
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.