Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Campylobacter Thioglycollate Medium with Five Antimicrobics

Catalog No. BB21747 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21747 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21747 Supplier BD Supplier No. 221747
This item is not returnable. View return policy

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

Holding medium for samples suspected to contain Campylobacter jejuni subsp. jejuni prior to inoculation of solid medium

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Color Light Beige
Content And Storage 2°C to 8°C
Form Highly Viscous
Manufacturing Origin United States
Media Type Campylobacter Thioglycollate Medium
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 1.6g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L, Amphotericin B 2mg/L, Cephalothin 15mg/L, Trimethoprim 5mg/L, Vancomycin 10mg/L, Polymyxin B 2500U
For Use With (Application) Samples suspected to contain Campylobacter jejuni subspecies jejuni prior to inoculation of solid medium
Packaging Type Tube
Quantity 10/Pk.

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.