Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Chopped Meat Glucose Broth, Pre-Reduced II (Huntgate Cap)

Catalog No. L97305 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, CMPH2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L97305 ISO/DIS, 11014, CMPH2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L97305 Supplier BD Supplier No. 297305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prepared tubed media for isolation and identification of microorganisms

  • BD Mycoflask™ sterile, disposable and ready-to-use BD BBL™ culture media are prepared in the special BD Mycoflask.
  • This variety of media helps isolate and cultivate tubercle bacilli and fungi.
TRUSTED_SUSTAINABILITY

Specifications

Broth Type Chopped Meat Glucose Broth, PR II (Huntgate Cap)
Cap Type Huntgate Cap
Certifications/Compliance ISO/DIS, 11014, CMPH2
Color Beige
Content And Storage 2 to 25°C
Detects Tubercle bacilli, Fungi
Form Liquid
Format Broth, Tube
Manufacturing Origin United States
Media Type Chopped Meat Glucose Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Fastidious Organisms
Test Type Microbial Identification
Formulation Chopped Meat Glucose Broth Culture Medium
For Use With (Application) Isolate and Cultivate Tubercle bacilli and Fungi
Includes 10 K-tubes
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.