Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Columbia CNA Agar with 5% Sheep Blood/MacConkey II, I Plate™ Prepared Media

Catalog No. L21601 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
For Use With (Application):
Gram positive microorganisms
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
L21601 100/Pk. Gram positive microorganisms
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21601 Supplier BD Supplier No. 221601
Add to Cart
Add to Cart
This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

Columbia CNA Agar with 5% Sheep Blood

  • Selective and differential medium used for isolation and differentiation of gram-positive microorganisms
Chocolate II Agar
  • Selective and differential medium for detection of coliform organisms and enteric pathogens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Red
Additive Colistin, Nalidixic Acid, 5% Sheep Blood
Detects Gram-positive Microorganisms, Coliform Organisms, Enteric Pathogens
Form Liquid
Format Biplate
Manufacturing Origin United States
Media Type Columbia CNA Agar with 5% Sheep Blood/MacConkey II Agar
Target Organism Class Gram-positive Organisms
Formulation Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L, Sheep Blood, defibrinated 5%
For Use With (Application) Gram positive microorganisms
Packaging Type Biplate
Quantity 100/Pk.
Weight (Metric) 4350 g
Protein Source Sheep
Source Sheep
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.