Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Cooked Meat Medium, 8mL

Catalog No. L21508 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Color:
Brown
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
L21508 Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21508 Supplier BD Supplier No. 221508
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

Used in cultivation of anaerobes, particularly pathogenic Clostridium species

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Cooked Meat Medium
Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF
Color Brown
Content And Storage 2 to 25°C
Form Homogenous Pellets
Manufacturing Origin United States
Media Type Cooked Meat Medium
Product Type Broth Tube
Sterility Sterile
Test Type Microbial Cultivation
Fill Volume 8 mL
Formulation Heart Tissue granules 98g/L, Peptic digest of Animal Tissue 20g/L, Dextrose 2g/L, Sodium Chloride 5g/L
For Use With (Application) Cultivation of anaerobes, pathogenic clostridia
Includes 100 Tubes
Incubation Conditions 35 ±2°C; Anaerobic Atmosphere
Incubation Time 40 to 48 hr.
No. per Pack 100
Packaging Type Tube
Quantity 8 mL
Protein Source Beef Heart, Proteose Peptone
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.