Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Cooked Meat Medium with Glucose, Hemin and Vitamin K

Catalog No. L95982 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
9 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L95982 9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L95982 Supplier BD Supplier No. 295982
Add to Cart
Add to Cart

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

General-purpose medium for cultivation of obligate anaerobes, specifically Clostridium spp.

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CMPH2, MCM9
Color Brown
Content And Storage 2 to 25°C
Form Homogenous Pellets
Manufacturing Origin United States
Media Type Cooked Meat Medium
Formulation Heart Tissue granules 98g/L, Peptic digest of Animal Tissue 20g/L, Dextrose 2g/L, Sodium Chloride 5g/L
For Use With (Application) Cultivation of obligate anaerobes, Clostridium species
No. per Pack 10
Packaging Type Tube
Quantity 9 mL
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.