Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Crystal™ ID Systems

Catalog No. B4345000 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Product Type:
Enteric/Nonfermenter Kit
Gram-positive ID Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B4345000 Enteric/Nonfermenter Kit
B4345240 Gram-positive ID Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B4345000 Supplier BD Supplier No. 245000
This item is not returnable. View return policy

Includes

Lids, bases, inoculum fluid and incubation trays sufficient for 20 determinations

Miniaturized microbial ID systems use modified conventional, fluorogenic and chromogenic substrates

Intended for identification of clinically significant aerobic and anaeorobic gram-negative and gram-positive bacterial pathogens isolated from human specimens. During incubation, the stackable 10-device trays save space and prevent incubator mess. Following incubation, distinct chromogenic and fluorogenic reactions generate a profile number, which is keyed directly into the BBL Crystal MIND Software installed on a laboratory computer.

  • Onscreen display identifies the organism, reports on epidemiology, and conveys other pertinent information
  • Easy-to-use systems require no pipetting or reagent addition
  • Culture colonies are suspended in Crystal Broth; then inoculum suspension is poured directly into the Crystal device
  • Wells fill automatically by rocking base
  • Lid is snapped onto the device for safety and handling ease

  • Enteric/Non-Fermenter (E/NF) Kit performs overnight identification of clinically significant aerobic gram-negative Enterobacteriaceae isolates, as well as frequently isolated glucose nonfermenting gram-negative bacilli from human origin
  • Anaerobe ID (ANR) Kit is a four-hour test that identifies clinically significant anaerobes. Separate databases for Schaedler Blood Agar, CDC Anaerobe Blood Agars are included for improved specificity and increased flexibility
  • Neisseria/Haemophilus (N/H) ID Kit identifies Neisseria, Haemophilus, Moraxella, Gardnerella vaginalis and other fastidious bacteria in a four-hour test
  • Gram-Positive (GP) Kit identifies 121 gram-positive bacteria isolated from clinical specimens including both cocci and bacilli in an overnight test
  • Rapid Gram-Positive (RGP) Kit identifies gram-positive bacteria isolated from clinical specimens including both cocci and bacilli in a four-hour test

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Solid
Product Type Enteric/Nonfermenter Kit
For Use With (Application) Identification of clinically significant aerobic gram negative Enterobacteriaceae isolates and isolated Glucose nonfermenting gram negative Bacilli from Human origin
Includes Lids, bases, inoculum fluid and incubation trays sufficient for 20 determinations
Quantity 20 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.