Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ CTA Medium

Catalog No. p-3239926 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Media Type:
CTA Culture Medium
Packaging Type:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Packaging Type
BB21632 CTA Culture Medium Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21632 Supplier BD Supplier No. 221632
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Simple, basic medium for the maintenance and detection of motility of a wide variety of microorganisms; addition of carboyhydrates allows use in the determination of fermentation reactions

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC
Color Red-Orange
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type CTA Culture Medium
Formulation L-Cystine 0.5g/L, Pancreatic digest of Casein 20g/L, Agar 2.5g/L, Sodium Chloride 5g/L, Sodium Sulfite 0.5g/L, Phenol red 0.017g/L
For Use With (Application) For maintaining the growth of microorganisms
Packaging Type Tube
Quantity 8 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.