Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ D/E Neutralizing Agar Isolator Pack Media - Rodac™ Plates

Catalog No. B292646 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B292646 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B292646 Supplier BD Supplier No. 292646
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure

Rodac (Replicate Organism Detection and Counting) plates for surface sampling.

To use:

  • Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
  • When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
  • Use the included inner bag to transport plates from your critical environment

Compliance

Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AAMI
Color Gray
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type D/E Neutralizing Agar
Sterility Sterile
For Use With (Application) Designed for isolator system
Includes Inner bag for transportation
Packaging Type Sterile Rodac™ Plate
Quantity 100/Pk.
Weight (Metric) 4080 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.