Learn More
Description
Beef Extract Powder is a meat extract dried into a powder. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins. Additionally, due to differences in the process, Beef Extract Powder has a different molecular weight profile than other beef extract products in our portfolio. Customers have used Beef Extract Powder in a variety of microbial fermentation cell lines including Clostridium sp., Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.
Features:
- An animal origin (AO) nutritional supplement
- A mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins
Applications of Beef Extract Powder include:
- Used in a variety of microbial fermentation cell lines
- Used to produce numerous animal health vaccines
Specifications
Specifications
| Certifications/Compliance | AOAC, COMPF, SMWW, ISO/DIS, 11014 |
| Content And Storage | Store at 2°C to 25°C in cool, dry conditions in well-sealed containers. |
| Additive | Beef Extract |
| Form | Powder |
| For Use With (Application) | Dehydrated culture media |
| Packaging Type | Bottle |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
