Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dermatophyte Test Medium (DTM) Modified with Chloramphenicol Slants

Catalog No. L99701 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L99701 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L99701 Supplier BD Supplier No. 299701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

Used for detection and presumptive identification of dermatophytes from clinical and veterinary specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, MCM9
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Dermatophyte Test Medium
Product Type Agar Slants
Sterility Sterile
Formulation Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 24g/L, Phenol red 0.2g/L, Cycloheximide 0.5g/L, gentamicin 0.1g/L, Chloramphenicol 0.1g/L
For Use With (Application) Detection and presumptive identification of dermatophytes
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.