Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Penicillinase (1,000,000 units/mL)

Catalog No. B11897 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Additive:
Penicillinase
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Additive
B11897 Penicillinase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11897 Supplier BD Supplier No. 211897
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivation of heterotrophic microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 1000000U/mL
Additive Penicillinase
Form Liquid
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Packaging Type Tube
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.