Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco Media Additives/Ingredients/Reagents: Phosphate Buffer

Catalog No. B11544 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Additive:
Phosphate Buffer APH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Additive
B11544 Phosphate Buffer APH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11544 Supplier BD Supplier No. 211544
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO/DIS, 11014
Additive Phosphate Buffer APH
Form Powder
For Use With (Application) Preparation of dilution blanks for use in the examination of Waters, dairy products, foods and other materials
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.