Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Rhamnose

Catalog No. DF0175152 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
25 g
100 g
For Use With (Application):
Dehydrated culture media
Industrial microbial testing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
DF0175-15-2 100 g Industrial microbial testing
DF0175-13-4 25 g Dehydrated culture media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DF0175-15-2 Supplier BD Supplier No. 217520
Add to Cart
Add to Cart
This item is not returnable. View return policy

L-rhamnose

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Additive Rhamnose
Form Powder
For Use With (Application) Industrial microbial testing
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.