Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Soluble Starch

Catalog No. df0178177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0178-17-7 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0178-17-7 Supplier BD Supplier No. 217820
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides Starch for hydrolysis, detoxification of metabolic byproducts and as a carbon source

  • Improves growth response
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2 to 25°C
Additive Soluble Starch
Form Powder
For Use With (Application) Provides Starch for hydrolysis, detoxification of metabolic byproducts and as a carbon source
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.