Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Middlebrook OADC Enrichment
GSA_VA

Catalog No. L12240 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L12240 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L12240 Supplier BD Supplier No. 212240
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in qualitative procedures for isolation and cultivation of mycobacteria. BD Difco™ BBL™ Middlebrook OADC Enrichment is a dehydrated culture media.

  • 100 mL per bottle (6/sp)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive Middlebrook OADC Enrichment
Form Viscous
For Use With (Application) Middlebrook 7H10 Agar
Packaging Type Bottle
Quantity 6 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.