Learn More
BD BBL™ Hemo ID Quad Plate, with Growth Factors
Catalog No. B97890X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 Pack
Catalog No. Quantity
B97890X 10 Pack
1 options
This item is not returnable. View return policy
Catalog No. B97890X Supplier BD Supplier No. 297890
This item is not returnable. View return policy
Separates various growth factors by media type. Isolates, differentiates and identifies Haemophilus species based on the X- and V-factor growth requirements and the hemolysis of horse blood.
  • Facilitates identification and eliminates the difficulties associated with inspecting and interpreting undivided colonies
  • After 24 hours of incubation, the Quad Plate will indicate the presence or absence of growth in specific quadrants, depending on X- and V-factor requirements for the various Haemophilus species
TRUSTED_SUSTAINABILITY
Specifications
Type Hemo ID Quad Plate
Includes Growth factors
Quantity 10 Pack