Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Kligler Iron Agar Slants Prepared Tubed and Mycoflask™ Media

Catalog No. BB20897 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB20897 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB20897 Supplier BD Supplier No. 220897
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

Used for diifferentiation of Enterobacteriaceae members on basis of their ability to ferment dextrose and lactose and to produce sulfides

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, ISO, MCM9
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Kligler Iron Agar
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Lactose 10g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Thiosulfate 0.5g/L, Agar 15g/L, Phenol red 0.025g/L
For Use With (Application) Differentiation of gram negative enteric bacilli
Packaging Type Tube
Quantity 100 Pack

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.