Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Media Phenol Red Mannitol Broth

Catalog No. B11527 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11527 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11527 Supplier BD Supplier No. 211527
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For determination of fermentation reactions in differentiation of microorganisms

  • Product type: Dehydrated Culture Media and Ingredients
  • Groupings: N through P Products
  • Latex free media
  • Used for differentiation of Bacillus and Salmonella
  • Dehydrated appearance: Fine, homogeneous, free of extraneous material
  • Prepared Appearance: Medium, orange-red to rose-red, clear to slightly hazy
  • Reaction of 2%, at 25°C, pH: 7.3 ± 0.2
  • Quantity: 500g, 1 Each

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BAM
Color Red
Form Powder
Manufacturing Origin United States
Media Type Phenol Red Mannitol Broth
Packaging Type Tube with Tight Cap
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.