Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Medium D/E Neutralizing Broth

Catalog No. B4398318 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, AOAC, BAM, USDA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
B4398318 ISO/DIS, 11014, AOAC, BAM, USDA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B4398318 Supplier BD Supplier No. 298318
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For neutralizing and testing of antiseptics and disinfectants according Engley and Dey procedure

  • For cultivating microorganisms
  • Used for environmental sampling to neutralize broad spectrum of antiseptic and disinfectant chemicals that helps in determining its bactericidal or bacteriostatic activity

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, USDA
Color Purple
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type D/E Neutralizing Broth
Formulation Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 10g/L, Sodium Bisulfite 2.5g/L, Sodium Thioglycollate 1g/L, Sodium Thiosulfate 6g/L, Lecithin 7g/L, Polysorbate 80 5g/L, Bromcresol Purple 0.02g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 0.1g/L
For Use With (Application) Neutralizing and testing of antiseptics and disinfectants according to the procedure of Engley and Dey.1
Packaging Type Tube
Quantity 9 mL

For laboratory use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.