Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Medium Herrold's Egg Yolk Agar: With Mycobactin J

Catalog No. B222233 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
w/Mycobactin J; 100/Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B222233 w/Mycobactin J; 100/Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B222233 Supplier BD Supplier No. 222233
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective isolation and differentiation of Mycobacterium paratuberculosis

  • Available with amphotericin, nalidixic Acid, vancomycin
  • Features “Slant Surface”, a widely used format that gives enough surface area for growth and visualization

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Yellow
Form Liquid
Manufacturing Origin United States
Media Type Herrold's Egg Yolk Agar with Amphotericin, Nalidixic Acid, Vancomycin, with Mycobactin J
For Use With (Application) Selective isolation and differentiation of Mycobacterium paratuberculosis
Packaging Type Slant
Quantity 100/Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.