Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Medium Herrold's Egg Yolk Agar: Without Mycobactin J

Catalog No. B222241 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Box
Media Type:
Herrold's EYA w/o Mycobactin J
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
B222241 100/Box Herrold's EYA w/o Mycobactin J
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B222241 Supplier BD Supplier No. 222241
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective isolation and differentiation of Mycobacterium paratuberculosis

  • Available with amphotericin, nalidixic acid, vancomycin
  • Features “Slant Surface”, a widely used format that gives enough surface area for growth and visualization

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Green
Country of Origin United States
Additive AmB, NA, VAN antibiotics
Detects Mycobacterium paratuberculosis
Form Solid
Format Slant, Tube
Manufacturing Origin United States
Media Type Herrold's EYA w/o Mycobactin J
Product Type Prepared Microbiology Plate
Test Type Microbial Differentiation
Formulation Pancreatic digest of Casein 9g/L, Beef extract 2.7g/L, Sodium Chloride 4g/L, Sodium Pyruvate 4.1g/L, Malachite green 0.1g/L, Amphotericin B 0.05g/L, Agar 15.3g/L, Nalidixic Acid 0.05g/L, Vancomycin 0.05g/L, Egg Yolks 100mL, Glycerol 27mL, Purified Water 873.0mL
For Use With (Application) Selective isolation and differentiation of Mycobacterium paratuberculosis
Includes AmB, NA, VAN antibiotics
Packaging Type Slant
Quantity 100/Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.