Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ MGIT™ Mycobacteria Growth Indicator Tube: Test Kits
GSA_VA

Catalog No. B4345113 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Tubes
25 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4345113 100 Tubes
B4345111 25 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B4345113 Supplier BD Supplier No. 245113
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers superior detection time in a user-friendly, fluorescent test format

  • Creates an environment suitable for rapid mycobacterial growth
  • Recommended for testing sputum, stool specimens, body fluids and tissue
  • The tube uses fluorometric technology to aid in the reading of results
  • Under UV light, a positive test emits a vivid orange fluorescent glow at the tube base and at the meniscus; a negative test shows little or no glow
  • The Mycobacteria can be removed from the tube for further testing
  • To use, supplement tube with MGIT OADC Enrichment and MGIT PANTA™ Antibiotic Mixture, inoculate with the prepared specimen and incubate
  • Tubes fit into standard procedures for decontamination and waste disposal

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Clear and Colorless
Broth Type Modified Middlebrook 7H9 Broth Base
Certifications/Compliance CE
Color Clear and Colorless
Content And Storage 2 to 25°C
Additive Fluorescent Indicator
Detects Mycobacteria
Format Test Kit, Tube
Manufacturing Origin United States
Media Type Mycobacteria Growth Indicator
Product Type Prepared Microbiology Tubes
Target Organism Class Mycobacteria
Test Type Microbial Detection
Fill Volume 4 mL
Formulation Modified Middlebrook 7H9 Broth base 110 μL fluorescent indicator containing Tris 4, 7-diphenyl-1,10-phenanthroline ruthenium chloride pentahydrate in a silicone rubber base
For Use With (Application) For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
Includes 100 Tubes
Incubation Conditions 37°C
Material Plastic Tube, Polypropylene Cap
No. per Pack 100
Packaging Type Carton
Quantity 100 Tubes
Dimensions (Dia. x H) 16 x 100 mm
Height (Metric) 100 mm
Diameter (Metric) 16 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.