Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Gruft Lowenstein-Jensen Media with Penicillin and Nalidixic Acid

Catalog No. L97653 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, BS12, CMPH2, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L97653 ISO/DIS, 11014, BS12, CMPH2, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L97653 Supplier BD Supplier No. 297653
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective media used for isolation and cultivation of Mycobacteria

  • Available as prepared slants (A Tubes)

TRUSTED_SUSTAINABILITY

Specifications

Appearance Beige Solid
Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2, MCM9
Color Beige
Content And Storage 2°C to 8°C
Additive Penicillin, Nalidixic Acid
Detects Tubercle bacilli, Fungi
Form Liquid
Format Tube, Slant
Manufacturing Origin United States
Media Type Lowenstein-Jensen Medium
Product Type Agar Slants
Sterility Sterile
Target Organism Class Mycobacteria
For Use With (Application) Isolate and Cultivate Tubercle bacilli and Fungi
No. per Pack 100
Packaging Type Tube with Tight Cap
Quantity 100 Pack
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.