Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Middlebrook 7H9 Broth with Glycerol Prepared Media

Catalog No. L21832 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21832 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21832 Supplier BD Supplier No. 221832
Add to Cart
Add to Cart
This item is not returnable. View return policy

For growth of pure cultures of mycobacteria for use in laboratory studies

  • Sterile, disposable, and ready-to-use culture media
  • Contains Glycerol
  • Helps isolate and cultivate tubercle bacilli and fungi
  • Used for qualitative procedures for the cultivation of mycobacteria
TRUSTED_SUSTAINABILITY

Specifications

Appearance Colorless to Very light Amber, Clear
Broth Type Middlebrook 7H9 Broth with Glycerol
Certifications/Compliance CE, AOAC, SMWW
Color Clear
Content And Storage 2°C to 8°C
Additive Glycerol
Detects Mycobacteria, Fungi, Tubercle bacilli
Form Liquid
Format Liquid
Manufacturing Origin United States
Media Type Middlebrook 7H9 Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Mycobacteria
Test Type Microbial Cultivation
Fill Volume 5 mL
Formulation Monopotassium Phosphate 2g/L, Disodium Phosphate 1.5g/L, Monosodium Glutamate 0.5g/L, Sodium Citrate 0.1g/L, Ammonium Sulfate 0.5g/L, Pyridoxine 0.001g/L, Ferric Ammonium Citrate 0.04g/L, Magnesium Sulfate 0.05g/L, Zinc Sulfate 0.001g/L, Copper Sulfate 0.001g/L, Biotin 0.5mg/L, Calcium Chloride 0.5mg/L
For Use With (Application) For growth of pure cultures of mycobacteria
Includes 10 Tubes
Incubation Time 21 days, 8 weeks
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 5 mL
Protein Source Bovine Albumin (Fraction V)
Show More Show Less

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.