Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Middlebrook and Cohn 7H10 Prepared Agar Slants

Catalog No. BB20959 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
Format:
Agar Slant
Packaging Type:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
BB20959 100 Pack Agar Slant Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB20959 Supplier BD Supplier No. 220959
Add to Cart
Add to Cart
This item is not returnable. View return policy

Prepared media. BBL™ Mycoflask™ Middlebrook and Cohn 7H10 Prepared Agar Slants are used in qualitative procedures for the isolation and cultivation of mycobacteria.

  • Contains a variety of inorganic salts that provide substances essential for the growth of mycobacteria
TRUSTED_SUSTAINABILITY

Specifications

Appearance Light Amber, Slightly Opalescent
Cap Type Screw Cap
Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2, EP, MCM9
Color Gray
Content And Storage 2°C to 8°C
Additive Sodium Chloride, Dextrose, Bovine Albumin, Catalase, Oleic Acid
Detects Mycobacteria
Form Highly Viscous
Format Agar Slant
Manufacturing Origin United States
Media Type Middlebrook and Cohn 7H10 Agar
Product Type Agar Slants
Sterility Sterile
Target Organism Class Mycobacteria
Test Type Microbial Cultivation
Formulation Magnesium Sulfate 0.05g/L, Ferric Ammonium Citrate 0.04g/L, Sodium Citrate 0.4g/L, Ammonium Sulfate 0.5g/L, Monosodium Glutamate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Agar 13.5g/L, Sodium Chloride 0.85g/L, Dextrose 2g/L, Bovine Albumin (Fraction V) 5g/L, Catalase 3mg/L, Pyridoxine 1mg/L, Zinc Sulfate 1mg/L, Copper Sulfate 1mg/L, Biotin 0.5mg/L, Calcium Chloride 0.5mg/L, Malachite green 0.25mg/L, Oleic Acid 0.06mLg/L, Glycerol 5mLg/L
For Use With (Application) Qualitative procedures for the isolation and cultivation of mycobacteria
Incubation Conditions CO2 Enriched Aerobic Atmosphere, 35 ± 2°C; Tubes in Slanted Position at 5° angle
No. per Pack 100
Packaging Type Tube
Quantity 100 Pack
Dimensions (Dia. x H) 20 x 148 mm
Protein Source Bovine
Show More Show Less

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.