Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Modified Trichosel™ Broth Prepare Media

Catalog No. L98323 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L98323 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L98323 Supplier BD Supplier No. 298323
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the isolation and cultivation of Trichomonas species

  • With 5% Horse Serum

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Light Beige Liquid
Broth Type Trichosel Broth, Modified, with 5% Horse Serum
Cap Type Tight Cap
Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Concentration 5% Horse Serum
Content And Storage 2°C to 8°C
Additive Chloramphenicol, Methylene Blue, Horse Serum
Detects Trichomonas Species
Form Liquid
Format Liquid, Tube
Manufacturing Origin United States
Media Type Trichosel Broth, Modified, with 5% Horse Serum
Product Type Broth Tube
Sterility Sterile
Test Type Microbial Cultivation
Fill Volume 10 mL
Formulation Pancreatic digest of Casein 12g/L, Beef extract 2g/L, Yeast extract 5g/L, L-Cysteine HCI 1g/L, Maltose 2g/L, Agar 1g/L, Chloramphenicol 0.1g/L, Methylene blue 3mg/L, Horse Serum 5%
For Use With (Application) Used for the isolation and cultivation of Trichomonas species
Includes Tubes
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 10 mL
Protein Source Casein, Beef, Yeast Extract, Horse Serum
Source Horse, Bovine, Yeast
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.