Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Motility Test Medium

Catalog No. BB21509 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21509 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BB21509 Supplier BD Supplier No. 221509
Add to Cart
Add to Cart

Prepared media used for the detection of motility of gram-negative enteric bacilli

TRUSTED_SUSTAINABILITY

Specifications

Cap Type Tight Cap
Certifications/Compliance CE, BAM, CCAM, COMPF, USDA
Color Beige
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Product Type Prepared Microbiology Tubes
Target Organism Class Gram-negative Rods
Test Type Microbial Detection
Formulation Beef extract 3g/L, Pancreatic digest of Gelatin 10g/L, Sodium Chloride 5g/L, Agar 4g/L
For Use With (Application) Detection of motility of gram negative enteric bacilli
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 10/Pk.
Height (Metric) 100 mm
Show More Show Less

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.