Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Mueller Hinton Broth Prepared Media

Catalog No. L96164 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, CLSI
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L96164 ISO/DIS, 11014, CLSI
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L96164 Supplier BD Supplier No. 296164
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in the cultivation of wide variety of fastidious and nonfastidious microorganisms

Also Available: BD BBL™ Mycoflask™ Mueller Hinton II Broth (Cation-Adjusted) Prepared Media (see 298268). 
TRUSTED_SUSTAINABILITY

Specifications

Broth Type Mueller Hinton Broth
Certifications/Compliance ISO/DIS, 11014, CLSI
Color Dark Yellow
Content And Storage 2 to 25°C
Detects Microorganisms, Tubercle bacilli, Fungi
Form Liquid
Format Liquid, K-tube
Manufacturing Origin United States
Media Type Mueller Hinton Broth
Formulation Acid Hydrolysate of Casein 17.5g/L, Beef extract 3g/L, Starch 1.5g/L
For Use With (Application) Microbial Cultivation
No. per Pack 100
Packaging Type K Tube with Tight Cap
Quantity 2 mL
Show More Show Less

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.