Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Mycobactosel™ L-J Medium Slants Prepared Media

Catalog No. BB21414 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, CMPH2, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
BB21414 ISO/DIS, 11014, CMPH2, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21414 Supplier BD Supplier No. 221414
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in qualitative procedures for the isolation of mycobacteria from clinical specimens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, CMPH2, MCM9
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Formulation Monopotassium Phosphate 2.5g/L, Magnesium Sulfate 0.24g/L, Sodium Citrate 0.6g/L, L-Asparagine 3.6g/L, Potato Flour 3g/L, Malachitegreen 0.4g/L, Cycloheximide 0.64g/L, Lincomycin 0.0032g/L, Nalidixic Acid 0.056g/L, Glycerol 12mL, Whole Egg 1L
For Use With (Application) Used for qualitative procedures for the isolation of mycobacteria from clinical specimens
Packaging Type Tube with Tight Cap
Quantity 100 Pack

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.