Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Mycosel™ Prepared Agar Slants

Catalog No. BB20967 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
Media Type:
Mycosel Agar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
BB20967 100 Pack Mycosel Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB20967 Supplier BD Supplier No. 220967
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria

  • Selective medium containing cycloheximide and chloramphenicol
  • For recovery of dermatophytes

Tubes, bottles and flasks with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2, MCM9
Color Light Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Mycosel Agar
Formulation Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 15.5g/L, Cycloheximide 0.4g/L, Chloramphenicol 0.05g/L
For Use With (Application) Isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria
Packaging Type Tube
Quantity 100 Pack

For in vitro Diagnostic Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.