Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Nutrient Agar Slants

Catalog No. BB20971 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB20971 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB20971 Supplier BD Supplier No. 220971
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivation of bacteria and enumeration of organisms in water, sewage, feces and other materials

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, ISO, SMWW, USDA
Color Beige
Detects Tubercle bacilli, Fungi
Form Powder
Format Agar Slant, Tube
Manufacturing Origin United States
Media Type Nutrient Agar
Product Type Agar Slants
Sterility Sterile
Target Organism Class Non-fastidious Organisms
Test Type Microbiological Tests
Formulation Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L, Agar 15g/L
For Use With (Application) For enumeration of organisms in Water, sewage, feces
No. per Pack 100
Packaging Type Tube with Tight Cap
Quantity 100 Pack
Show More Show Less

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.