Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Nutrient Broth Prepared Media

Catalog No. BB21669 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21669 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BB21669 Supplier BD Supplier No. 221669
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For cultivation of many species of nonfastidious microorganisms

Nutrient Broth has the formula originally designed for use in the standard methods for the examination of water and wastewater.

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Cap Type Tight Cap
Certifications/Compliance CE, BAM, CCAM, COMPF, SMD
Color Beige
Detects Non-fastidious Microorganisms, Tubercle bacilli, Fungi
Form Powder
Format Liquid, Pour Tube
Manufacturing Origin United States
Media Type Nutrient Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Non-fastidious Organisms
Fill Volume 5 mL
Formulation Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L
For Use With (Application) Cultivation of many species of non-fastidious microorganisms
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 5 mL
Protein Source Beef, Gelatin
Source Bovine
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.