Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ OF (Oxidation Fermentation) Media

Catalog No. BB21328 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Media Type:
OF media with Dextrose
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
BB21328 OF media with Dextrose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21328 Supplier BD Supplier No. 221328
Add to Cart
Add to Cart
This item is not returnable. View return policy

Determines oxidative and fermentative metabolism of carbohydrates by Gram-negative rods on the basis of acid reaction

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Blue
Content And Storage 2°C to 8°C
Additive Dextrose
Detects Oxidative and Fermentative Metabolism
Form Liquid
Manufacturing Origin United States
Media Type OF media with Dextrose
Sterility Sterile
Target Organism Class Gram-negative Bacteria
Test Type Presumptive Identification
Formulation Pancreatic digest of Casein 2g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 0.3g/L, Agar 2.5g/L, Bromthymol blue 0.03g/L
For Use With (Application) Determination of oxidative and fermentative metabolism of carbohydrates by gram negative rods on the basis of Acid reaction
Packaging Type Tube
Quantity 10/Pk.
Protein Source Casein
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.