Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Phenol Red Broth Prepared Media with Dextrose and Durham Tube

Catalog No. BB21677 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CE, BAM
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
BB21677 CE, BAM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21677 Supplier BD Supplier No. 221677
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for determination of fermentation reactions in the differentiation of microorganisms

The ability of an organism to ferment dextrose incorporated in a basal medium, resulting in the production of acid or acid and gas, has been used to characterize a specific species or group of bacteria, aid in the differentiation between genera, and aid in species differentiation.

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Color Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Phenol Red Broth
Formulation Pancreatic digest of Casein 10g/L, Sodium Chloride 5g/L, Phenol red 0.018g/L
For Use With (Application) Determination of fermentation reactions in the differentiation of microorganisms
Packaging Type Tube with Tight Cap
Quantity 10/Pk.

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.