Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Phosphate Buffer, pH 6.8, 500mL
GSA_VA

Catalog No. L97657 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L97657 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L97657 Supplier BD Supplier No. 297657
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used as buffered diluent for microbiological procedures

For use with Mycoflask Phosphate Buffer

  • Volume: 500mL
  • Color: Beige

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Clear and Colorless
Certifications/Compliance ISO/DIS, 11014
Color Beige
Form Liquid
Format Liquid
Manufacturing Origin United States
Sterility Sterile
Target Organism Class Non-fastidious Organisms
Test Type Microbiological Tests
Fill Volume 500 mL
Formulation Phosphate Buffer
For Use With (Application) Used for a Buffered diluent for microbiological procedures
Incubation Conditions 20°C to 25°C, 33°C to 37°C
Incubation Time 10 min, up to 3 days
No. per Pack 1
Packaging Type Bottle
Quantity 500 mL
Manufacturing Quality ISO 9001:2008, ISO 13485:2003, FDA-registered
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.