Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Phosphate Buffer, pH 6.8, 600mL, P80

Catalog No. B215095 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B215095 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B215095 Supplier BD Supplier No. 215095
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used as a buffered diluent for microbiological procedures

For use with Mycoflask Phosphate Buffer

  • Volume: 600mL
  • P80
  • Colorless

TRUSTED_SUSTAINABILITY

Specifications

Product Type Buffer Solution
Fill Volume 600 mL
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.