Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Polysorbate 80 Water

Catalog No. L96184 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L96184 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L96184 Supplier BD Supplier No. 296184
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used to restore and/or inoculate microdilution panels

Care should be exercised in opening tubes with tight caps to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Polysorbate 80 Water
Formulation Polysorbate 80 0.2mL
For Use With (Application) Used for restore/inoculate microdilution panels, recommended for use at 0.02% concentration in routine inoculum preparation and dispensing procedures
Packaging Type Tube with Tight Cap
Quantity 25 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.