Learn More
BD BBL™ Mycoflask™ Prepared Bottled Media, Standard Methods Agar
Shop All BD Diagnostic Systems ProductsDescription
- Ready-to-use formulations for detecting microorganisms from pharmaceutical, food, environmental, or water testing samples.
- Offered in various bottle styles and fill volumes to meet customer requirements.
- Prepared using highest quality BBL and Difco brand dehydrated culture media.
- Conforms to current USP and EP requirements (see package labeling for details).
- Color-coded caps and/or labeling for most products, providing easy product identification.
- Transparent labeling to allow a clear view of media and culture.
Specifications
Specifications
| Cap Type | Screw Cap |
| Detects | Microorganisms |
| Form | Prepared Medium |
| Format | Bottle |
| Media Type | Standard Methods Agar |
| Preparation Method | Prepared |
| Product Type | Culture Medium |
| Test Type | Environmental Test |
| Fill Volume | 500 mL |
| For Use With (Application) | Detecting Microorganisms, Food, Water Testing Samples |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.