Learn More
BD BBL™ Mycoflask™ Prepared Lauryl Sulfate Broth with Durham Tube, 10mL
Shop All BD Diagnostic Systems ProductsSpecifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2 to 25°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Lauryl Tryptose Broth |
| Formulation | Sodium Lauryl Sulfate 0.2g/L, Pancreatic digest of Casein 40g/L, Lactose 10g/L, Dipotassium Phosphate 5.5g/L, Monopotassium Phosphate 5.5g/L, Sodium Chloride 10g/L |
| For Use With (Application) | Detection of Coliform organisms in materials of sanitary importance |
| Packaging Type | Tube with Tight Cap |
| Quantity | 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.