Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Lauryl Sulfate Broth with Durham Tube, 10mL

Catalog No. p-4639631 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Media Type:
Lauryl Tryptose Broth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
BB21248 Lauryl Tryptose Broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21248 Supplier BD Supplier No. 221248
This item is not returnable. View return policy

For the detection of coliform organisms in materials of sanitary importance

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Lauryl Tryptose Broth
Formulation Sodium Lauryl Sulfate 0.2g/L, Pancreatic digest of Casein 40g/L, Lactose 10g/L, Dipotassium Phosphate 5.5g/L, Monopotassium Phosphate 5.5g/L, Sodium Chloride 10g/L
For Use With (Application) Detection of Coliform organisms in materials of sanitary importance
Packaging Type Tube with Tight Cap
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.