Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Lim Broth (Todd Hewitt with CNA) Media

Catalog No. L96266 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
5 mL
Color:
Beige
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
L96266 5 mL Beige
L4392209 10/Pk. Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L96266 Supplier BD Supplier No. 296266
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For selective enrichment of group B Streptococci (Streptococcus agalactiae), especially from genital specimens

  • Available as K tubes

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CMPH2, MCM9
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Lim Broth
Formulation Heart infusion from (solids) 3.1g/L, Peptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L, Yeast extract 10g/L, Colistin 0.01g/L, Nalidixic Acid 0.015g/L
For Use With (Application) For selective enrichment of group B Streptococci, especially from genital specimens
Packaging Type Tube
Quantity 5 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.